Product Details

SNP ID
rs80301407
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:35238154 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGATAACTGCAGAGCCGAGGCACA[A/G]AGAGCAGAGGCTGGCTACTGAGCCA
Phenotype
MIM: 604871
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MMP24 PubMed Links

Gene Details

Gene
MMP24
Gene Name
matrix metallopeptidase 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006690.3 Intron NP_006681.1
XM_011528500.2 Intron XP_011526802.1
XM_017027597.1 Intron XP_016883086.1
XM_017027598.1 Intron XP_016883087.1

View Full Product Details