Product Details

SNP ID
rs77000936
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:23633940 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTAGGCGTCCTGACAGGTGGATTT[C/T]GACAAGGTCATTGTGCCCTGCCAAG
Phenotype
MIM: 604312
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CST3 PubMed Links

Gene Details

Gene
CST3
Gene Name
cystatin C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000099.3 603 Silent Mutation TCA,TCG S139S NP_000090.1
NM_001288614.1 603 Silent Mutation TCA,TCG S139S NP_001275543.1

View Full Product Details