Product Details

SNP ID
rs74877410
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:33168633 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCCAAGTAACAGCAATGGGGGCT[A/G]GGAGGAGTGAAAGAGGGAGGTGGGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC01548 PubMed Links

Gene Details

Gene
LINC01548
Gene Name
long intergenic non-protein coding RNA 1548
There are no transcripts associated with this gene.

View Full Product Details