Product Details

SNP ID
rs76299166
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:34673998 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTGTGGAATTCACAGCTGGGTTT[A/G]AAGGAAAGTACTGCAATATTCTCTC
Phenotype
MIM: 615321
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLIC6 PubMed Links

Gene Details

Gene
CLIC6
Gene Name
chloride intracellular channel 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317009.1 Intron NP_001303938.1
NM_053277.2 Intron NP_444507.1
XM_017028406.1 Intron XP_016883895.1

View Full Product Details