Product Details

SNP ID
rs80298637
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.21:41424602 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAGCTAAGGAGAATGGAGGTGGG[G/T]TGCCAACCGCTCCCAAAGGGAAATG
Phenotype
MIM: 147150
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
MX1 PubMed Links

Gene Details

Gene
MX1
Gene Name
MX dynamin like GTPase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144925.2 Intron NP_001138397.1
NM_001178046.2 Intron NP_001171517.1
NM_001282920.1 Intron NP_001269849.1
NM_002462.4 Intron NP_002453.2
XM_005260978.4 Intron XP_005261035.1
XM_005260979.2 Intron XP_005261036.1
XM_005260980.2 Intron XP_005261037.1
XM_005260981.2 Intron XP_005261038.1
XM_005260982.2 Intron XP_005261039.1
XM_011529568.2 Intron XP_011527870.1
XM_011529570.2 Intron XP_011527872.1
XM_017028349.1 Intron XP_016883838.1
XM_017028350.1 Intron XP_016883839.1
XM_017028351.1 Intron XP_016883840.1
XM_017028352.1 Intron XP_016883841.1

View Full Product Details