Product Details
- SNP ID
-
rs79395473
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:46291430 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTGGAGTATATCTTCCATCAGTGT[A/G]AAGAAAATGAAGATTACAATGACGT
- Phenotype
-
MIM: 603294
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
C21orf58
PubMed Links
Gene Details
- Gene
- C21orf58
- Gene Name
- chromosome 21 open reading frame 58
There are no transcripts associated with this gene.
- Gene
- MCM3AP
- Gene Name
- minichromosome maintenance complex component 3 associated protein
There are no transcripts associated with this gene.
- Gene
- YBEY
- Gene Name
- ybeY metallopeptidase (putative)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001006114.2 |
407 |
Missense Mutation |
AAA,GAA |
K103E |
NP_001006114.1 |
NM_001314022.1 |
407 |
Intron |
|
|
NP_001300951.1 |
NM_001314023.1 |
407 |
Missense Mutation |
AAA,GAA |
K58E |
NP_001300952.1 |
NM_001314024.1 |
407 |
Intron |
|
|
NP_001300953.1 |
NM_001314025.1 |
407 |
Missense Mutation |
AAA,GAA |
K103E |
NP_001300954.1 |
NM_001314026.1 |
407 |
Missense Mutation |
AAA,GAA |
K103E |
NP_001300955.1 |
NM_058181.2 |
407 |
Missense Mutation |
AAA,GAA |
K103E |
NP_478061.1 |
XM_005261156.3 |
407 |
Intron |
|
|
XP_005261213.2 |
XM_011529629.2 |
407 |
Missense Mutation |
AAA,GAA |
K103E |
XP_011527931.1 |
XM_011529630.2 |
407 |
Missense Mutation |
AAA,GAA |
K103E |
XP_011527932.1 |
XM_011529631.2 |
407 |
Missense Mutation |
AAA,GAA |
K103E |
XP_011527933.1 |
XM_011529633.2 |
407 |
Missense Mutation |
AAA,GAA |
K103E |
XP_011527935.1 |
XM_011529634.2 |
407 |
Intron |
|
|
XP_011527936.2 |
XM_011529635.2 |
407 |
Missense Mutation |
AAA,GAA |
K103E |
XP_011527937.1 |
XM_017028394.1 |
407 |
Intron |
|
|
XP_016883883.1 |
XM_017028395.1 |
407 |
Intron |
|
|
XP_016883884.1 |
XM_017028396.1 |
407 |
Intron |
|
|
XP_016883885.1 |
View Full Product Details