Product Details

SNP ID
rs78099126
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:36762941 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAACATTCCAGGCCCTGGCCCAGC[A/G]CCCATGCCCGGGCCTCAGCTGAGTC
Phenotype
MIM: 615870
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IFT27 PubMed Links

Gene Details

Gene
IFT27
Gene Name
intraflagellar transport 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177701.2 855 Missense Mutation GCG,GTG A142V NP_001171172.1
NM_006860.4 855 Missense Mutation GCG,GTG A141V NP_006851.1
XM_006724106.2 855 Missense Mutation GCG,GTG A142V XP_006724169.1
XM_017028540.1 855 Missense Mutation GCG,GTG A101V XP_016884029.1
Gene
LOC105373021
Gene Name
uncharacterized LOC105373021
There are no transcripts associated with this gene.

View Full Product Details