Product Details

SNP ID
rs74525416
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.22:21923112 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCACCTAGCTTCTTGGTGGAGCCT[C/G]GGTCTCAGGTTCTGGAAGGCTGGAG
Phenotype
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
PPM1F PubMed Links

Gene Details

Gene
PPM1F
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014634.3 1492 Missense Mutation CAG,GAG Q449E NP_055449.1
XM_011530556.1 1492 Missense Mutation CAG,GAG Q281E XP_011528858.1
XM_017029108.1 1492 Intron XP_016884597.1

View Full Product Details