Product Details

SNP ID
rs79294516
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:26433057 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGACATGGGCCTTGAAGTCAGAT[A/G]GGGTTTGAGTCTCTGCCCTGGCTGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASPHD2 PubMed Links

Gene Details

Gene
ASPHD2
Gene Name
aspartate beta-hydroxylase domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020437.4 Intron NP_065170.2

View Full Product Details