Product Details

SNP ID
rs77411392
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:101837514 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCTCTGGCTCTCTGGGCCTGCCC[A/G]AGGGAGGAAGCAGGTGTTCACGGTC
Phenotype
MIM: 300319
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NXF5 PubMed Links

Gene Details

Gene
NXF5
Gene Name
nuclear RNA export factor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032946.2 1420 Missense Mutation CGG,TGG R354W NP_116564.2

View Full Product Details