Product Details

SNP ID
rs78737354
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:76215705 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGCTCTTGGAGCCTCAAGTCACC[C/T]CTAAGTTCAGCTACCCGGAGACTGC
Phenotype
MIM: 602257
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM47E PubMed Links

Gene Details

Gene
FAM47E
Gene Name
family with sequence similarity 47 member E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136570.2 Intron NP_001130042.1
NM_001242936.1 Intron NP_001229865.1
Gene
SCARB2
Gene Name
scavenger receptor class B member 2
There are no transcripts associated with this gene.

View Full Product Details