Product Details
- SNP ID
-
rs76844320
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
12
- Location
-
Chr.1:9162714 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCCCTACCCTTCAATAAGGATCAC[A/C]CAGCACTTGGGGTCCTATCTGGCTT
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MIR34AHG
PubMed Links
Gene Details
- Gene
- MIR34AHG
- Gene Name
- MIR34A host gene
There are no transcripts associated with this gene.
View Full Product Details