Product Details

SNP ID
rs79995992
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:9924971 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGGCTTCAAAATTCAGTTACAGT[C/T]GCCAGAGGCTGAGGCAGAGGATCTG
Phenotype
MIM: 610458
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LZIC PubMed Links

Gene Details

Gene
LZIC
Gene Name
leucine zipper and CTNNBIP1 domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316973.1 Intron NP_001303902.1
NM_001316974.1 Intron NP_001303903.1
NM_001316975.1 Intron NP_001303904.1
NM_001316976.1 Intron NP_001303905.1
NM_032368.4 Intron NP_115744.2
XM_005263506.2 Intron XP_005263563.1
XM_005263509.2 Intron XP_005263566.1
XM_017002546.1 Intron XP_016858035.1
XM_017002547.1 Intron XP_016858036.1

View Full Product Details