Product Details

SNP ID
rs77982861
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:10011045 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAGACAGAAAACAAAGAAATTTC[A/G]CAGGATTACAACATTTTCTATACAA
Phenotype
MIM: 608604
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RBP7 PubMed Links

Gene Details

Gene
RBP7
Gene Name
retinol binding protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052960.2 Intron NP_443192.1

View Full Product Details