Product Details

SNP ID
rs74511329
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:10220485 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTTCTTTAAGTAAAAGGAAGAA[A/C]TGTTCTTCAAATACCTTTACAAAAT
Phenotype
MIM: 605995
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KIF1B PubMed Links

Gene Details

Gene
KIF1B
Gene Name
kinesin family member 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015074.3 Intron NP_055889.2
NM_183416.3 Intron NP_904325.2
Gene
MIR1273D
Gene Name
microRNA 1273d
There are no transcripts associated with this gene.

View Full Product Details