Product Details

SNP ID
rs77112774
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:24945412 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTCTCTGCTGTAATTGCTCATCT[G/T]TGCAATACGGCTGAATGAGGTTCCC
Phenotype
MIM: 613527
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DNAJC27 PubMed Links

Gene Details

Gene
DNAJC27
Gene Name
DnaJ heat shock protein family (Hsp40) member C27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198559.1 3107 UTR 3 NP_001185488.1
NM_016544.2 3107 UTR 3 NP_057628.1
XM_011532902.1 3107 Intron XP_011531204.1
XM_017004269.1 3107 UTR 3 XP_016859758.1
XM_017004270.1 3107 UTR 3 XP_016859759.1

View Full Product Details