Product Details

SNP ID
rs80108312
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:32482263 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAATCCTAAAGGTATTGAGTCAG[C/T]AAGACAGAGCAGAAGCTATGTTGGG
Phenotype
MIM: 607889
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CMTM6 PubMed Links

Gene Details

Gene
CMTM6
Gene Name
CKLF like MARVEL transmembrane domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017801.2 2415 UTR 3 NP_060271.1

View Full Product Details