Product Details

SNP ID
rs74438730
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:51567800 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCTAAGTAGAGGCCTTTTCAGTA[C/T]GTCATATCCAAGCTAGCACAAAAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAD54L2 PubMed Links

Gene Details

Gene
RAD54L2
Gene Name
RAD54-like 2 (S. cerevisiae)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322253.1 Intron NP_001309182.1
NM_001322256.1 Intron NP_001309185.1
NM_001322257.1 Intron NP_001309186.1
NM_015106.3 Intron NP_055921.2
XM_006713062.3 Intron XP_006713125.1
XM_017005987.1 Intron XP_016861476.1
XM_017005988.1 Intron XP_016861477.1

View Full Product Details