Product Details

SNP ID
rs74762885
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:52322110 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGGGGGAGAGGGGTGGACAAGT[A/G]CAGTCGCATGCATGGGTTGGGGGCA
Phenotype
MIM: 603332
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAH1 PubMed Links

Gene Details

Gene
DNAH1
Gene Name
dynein axonemal heavy chain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015512.4 Intron NP_056327.4
XM_017006129.1 Intron XP_016861618.1
XM_017006130.1 Intron XP_016861619.1
XM_017006131.1 Intron XP_016861620.1
XM_017006132.1 Intron XP_016861621.1
XM_017006133.1 Intron XP_016861622.1

View Full Product Details