Product Details

SNP ID
rs74506626
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:70441126 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGTGGTATTACATGTATTTATA[G/T]GTTCTGGAAATTGAGTAGAGAAGCA
Phenotype
MIM: 158375
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MUC7 PubMed Links

Gene Details

Gene
MUC7
Gene Name
mucin 7, secreted
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145006.1 Intron NP_001138478.1
NM_001145007.1 Intron NP_001138479.1
NM_152291.2 Intron NP_689504.2

View Full Product Details