Product Details

SNP ID
rs75938900
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:70655643 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTTACAAAATGGATTGTTTCAAA[G/T]GGCAATTTCTTACACTAACCTGATT
Phenotype
MIM: 606585 MIM: 147790
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ENAM PubMed Links

Gene Details

Gene
ENAM
Gene Name
enamelin
There are no transcripts associated with this gene.

Gene
JCHAIN
Gene Name
joining chain of multimeric IgA and IgM
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144646.3 1209 UTR 3 NP_653247.1
XM_006714211.2 1209 UTR 3 XP_006714274.1
XM_006714212.2 1209 UTR 3 XP_006714275.1
XM_011531925.1 1209 UTR 3 XP_011530227.1
XM_011531926.1 1209 UTR 3 XP_011530228.1

View Full Product Details