Product Details

SNP ID
rs76674380
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:56395490 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTTTCTGTTTTTTAAACTTTAT[A/C]CCTTCTTGTACAGATGAAACCAGTC
Phenotype
MIM: 614365 MIM: 172450
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
AASDH PubMed Links

Gene Details

Gene
AASDH
Gene Name
aminoadipate-semialdehyde dehydrogenase
There are no transcripts associated with this gene.

Gene
PPAT
Gene Name
phosphoribosyl pyrophosphate amidotransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002703.4 Intron NP_002694.3
XM_011534387.1 Intron XP_011532689.1

View Full Product Details