Product Details

SNP ID
rs77874259
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136984760 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGCCTTCTCCGACTACCAGCAC[G/T]TTGCCTTGCTGTACTTGGAGATGCG
Phenotype
MIM: 616315 MIM: 606533 MIM: 176803
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C9orf142 PubMed Links

Gene Details

Gene
C9orf142
Gene Name
chromosome 9 open reading frame 142
There are no transcripts associated with this gene.

Gene
CLIC3
Gene Name
chloride intracellular channel 3
There are no transcripts associated with this gene.

Gene
LCNL1
Gene Name
lipocalin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207510.3 838 Missense Mutation GTT,TTT V82F NP_997393.3
Gene
PTGDS
Gene Name
prostaglandin D2 synthase
There are no transcripts associated with this gene.

View Full Product Details