Product Details

SNP ID
rs77753969
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:107039784 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATGGCTATGGCTAGTGTTAAATT[G/T]CTTGCCGGTGTTTTAAGAAAGCCAG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C6orf203 PubMed Links

Gene Details

Gene
C6orf203
Gene Name
chromosome 6 open reading frame 203
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142468.2 240 Missense Mutation TTG,TTT L8F NP_001135940.1
NM_001142470.2 240 Missense Mutation TTG,TTT L13F NP_001135942.1
NM_016487.4 240 Missense Mutation TTG,TTT L8F NP_057571.1
XM_011535876.2 240 Missense Mutation TTG,TTT L13F XP_011534178.1
XM_011535877.2 240 Missense Mutation TTG,TTT L13F XP_011534179.1
XM_017010919.1 240 Intron XP_016866408.1
XM_017010920.1 240 Missense Mutation TTG,TTT L38F XP_016866409.1

View Full Product Details