Product Details

SNP ID
rs78733835
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:133889838 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATACGAGAACGGGTACATTCACCC[G/T]GTCAACCTGGTGAGTGCTCCCGGGG
Phenotype
MIM: 616058 MIM: 603306
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
TARID PubMed Links

Gene Details

Gene
TARID
Gene Name
TCF21 antisense RNA inducing promoter demethylation
There are no transcripts associated with this gene.

Gene
TCF21
Gene Name
transcription factor 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003206.3 718 Silent Mutation CCG,CCT P147P NP_003197.2
NM_198392.2 718 Silent Mutation CCG,CCT P147P NP_938206.1

View Full Product Details