Product Details

SNP ID
rs80104385
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:138218298 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGCTAGAATGCCTGGTAGCCTT[G/T]GGAAGTTACTTAATCTGTGCCTCAA
Phenotype
MIM: 605669
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ARFGEF3 PubMed Links

Gene Details

Gene
ARFGEF3
Gene Name
ARFGEF family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020340.4 193 Intron NP_065073.3
Gene
PBOV1
Gene Name
prostate and breast cancer overexpressed 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021635.2 193 Missense Mutation CAA,CCA Q33P NP_067648.1

View Full Product Details