Product Details

SNP ID
rs78608409
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:143885363 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCCCCTGAGAATTAGTTTGAGAC[A/C]TGGGAGGTTCCATTTGCAGAGGGAT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZC2HC1B PubMed Links

Gene Details

Gene
ZC2HC1B
Gene Name
zinc finger C2HC-type containing 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013623.2 Intron NP_001013645.1

View Full Product Details