Product Details

SNP ID
rs77926621
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:161348847 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGGGTAAGAGCATGCGGTTTGCC[A/G]CATGCAGTGTTGTTAATCTTTTGAT
Phenotype
MIM: 602544
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PARK2 PubMed Links

Gene Details

Gene
PARK2
Gene Name
parkin RBR E3 ubiquitin protein ligase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004562.2 2781 UTR 3 NP_004553.2
NM_013987.2 2781 UTR 3 NP_054642.2
NM_013988.2 2781 UTR 3 NP_054643.2
XM_011535863.1 2781 UTR 3 XP_011534165.1
XM_017010908.1 2781 UTR 3 XP_016866397.1
XM_017010909.1 2781 UTR 3 XP_016866398.1

View Full Product Details