Product Details

SNP ID
rs76510347
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:166324424 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCCCTCCTCTGACCCTGCAGGG[C/T]CTGTCTGGCTCCAAAACACCAGACG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SFT2D1 PubMed Links

Gene Details

Gene
SFT2D1
Gene Name
SFT2 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145169.2 Intron NP_660152.1

View Full Product Details