Product Details

SNP ID
rs77163984
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:105160124 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCATATTATACTTCAATAAAACAT[C/T]TTAATGGACTTTGTCAGATTTTAAA
Phenotype
MIM: 605824
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BVES-AS1 PubMed Links

Gene Details

Gene
BVES-AS1
Gene Name
BVES antisense RNA 1
There are no transcripts associated with this gene.

Gene
POPDC3
Gene Name
popeye domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022361.4 Intron NP_071756.2
XM_011536067.2 Intron XP_011534369.1
XM_017011194.1 Intron XP_016866683.1
XM_017011195.1 Intron XP_016866684.1

View Full Product Details