Product Details

SNP ID
rs76309235
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:31451463 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCCTGGCCTCTCTCTCTCTCTCT[C/T]TTTTTTTCTTTCTTCTTCAGACCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DTD2 PubMed Links

Gene Details

Gene
DTD2
Gene Name
D-tyrosyl-tRNA deacylase 2 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080664.2 Intron NP_542395.1
Gene
LOC101927124
Gene Name
uncharacterized LOC101927124
There are no transcripts associated with this gene.

View Full Product Details