Product Details

SNP ID
rs76669080
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:58200252 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACGAGGGCCTGGGGAGCGGCGGGG[G/A]GAAGACGGCGGTCGTGATCGACCTG
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
ACTR10 PubMed Links

Gene Details

Gene
ACTR10
Gene Name
actin-related protein 10 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018477.2 94 Missense Mutation GAG,GGG E12G NP_060947.1
XM_011536960.1 94 Missense Mutation GAG,GGG E12G XP_011535262.1
XM_011536961.1 94 Missense Mutation GAG,GGG E12G XP_011535263.1
XM_011536962.2 94 UTR 5 XP_011535264.1
XM_011536963.2 94 UTR 5 XP_011535265.1

View Full Product Details