Product Details

SNP ID
rs74345106
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24631185 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGACAATGCCCTGGGCCACCTT[G/T]TTACACACAAGAGGGCCTCCAGAGT
Phenotype
MIM: 123910
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GZMB PubMed Links

Gene Details

Gene
GZMB
Gene Name
granzyme B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004131.4 680 Missense Mutation AAA,AAC K210N NP_004122.2
XM_011536685.2 680 Missense Mutation AAA,AAC K198N XP_011534987.1

View Full Product Details