Product Details

SNP ID
rs80340345
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:97926729 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTCAGGGAATGTTCCTCGCATAA[C/T]GAATGTTCATTGTGTATGATTTGTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC01550 PubMed Links

Gene Details

Gene
LINC01550
Gene Name
long intergenic non-protein coding RNA 1550
There are no transcripts associated with this gene.

View Full Product Details