Product Details

SNP ID
rs77658190
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:100834506 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGAGGCACCGGCAGAGGGGTCTC[A/G]TTGCTCCCTTGAGTGTAGGGGCAGC
Phenotype
MIM: 605636
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MEG3 PubMed Links

Gene Details

Gene
MEG3
Gene Name
maternally expressed 3 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details