Product Details

SNP ID
rs74474835
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:25337841 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTAACAACATCACATACACAGAAG[A/C]CTAGGAAAGGGGAAACTACTTACTT
Phenotype
MIM: 601623
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
UBE3A PubMed Links

Gene Details

Gene
UBE3A
Gene Name
ubiquitin protein ligase E3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000462.3 4337 UTR 3 NP_000453.2
NM_130838.1 4337 UTR 3 NP_570853.1
NM_130839.2 4337 UTR 3 NP_570854.1
XM_005268267.4 4337 UTR 3 XP_005268324.1
XM_005268268.4 4337 UTR 3 XP_005268325.1
XM_005268269.4 4337 UTR 3 XP_005268326.1
XM_005268270.4 4337 UTR 3 XP_005268327.1
XM_005268271.4 4337 UTR 3 XP_005268328.1
XM_006720675.3 4337 UTR 3 XP_006720738.1
XM_006720676.3 4337 UTR 3 XP_006720739.1
XM_011521994.2 4337 UTR 3 XP_011520296.1
XM_011521995.2 4337 UTR 3 XP_011520297.1
XM_017022544.1 4337 UTR 3 XP_016878033.1
XM_017022545.1 4337 UTR 3 XP_016878034.1
XM_017022546.1 4337 UTR 3 XP_016878035.1
XM_017022547.1 4337 UTR 3 XP_016878036.1
XM_017022548.1 4337 UTR 3 XP_016878037.1
XM_017022549.1 4337 UTR 3 XP_016878038.1
XM_017022550.1 4337 UTR 3 XP_016878039.1
XM_017022551.1 4337 UTR 3 XP_016878040.1
XM_017022552.1 4337 UTR 3 XP_016878041.1
XM_017022553.1 4337 UTR 3 XP_016878042.1
XM_017022554.1 4337 UTR 3 XP_016878043.1
XM_017022555.1 4337 UTR 3 XP_016878044.1
XM_017022556.1 4337 UTR 3 XP_016878045.1
XM_017022557.1 4337 UTR 3 XP_016878046.1
XM_017022558.1 4337 UTR 3 XP_016878047.1
XM_017022559.1 4337 UTR 3 XP_016878048.1

View Full Product Details