Product Details

SNP ID
rs79142878
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:150896396 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTTGATGTACAATGAGTTGTGA[C/T]TTCTTAGCAAAGGCTTTTCCACATG
Phenotype
MIM: 608212 MIM: 612429
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IRGM PubMed Links

Gene Details

Gene
IRGM
Gene Name
immunity related GTPase M
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145805.1 1264 Intron NP_001139277.1
XM_011537641.2 1264 Intron XP_011535943.1
Gene
ZNF300
Gene Name
zinc finger protein 300
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172831.1 1264 Silent Mutation AAA,AAG K297K NP_001166302.1
NM_001172832.1 1264 Silent Mutation AAA,AAG K245K NP_001166303.1
NM_052860.2 1264 Silent Mutation AAA,AAG K281K NP_443092.1
XM_011537702.2 1264 Silent Mutation AAA,AAG K286K XP_011536004.1

View Full Product Details