Product Details

SNP ID
rs77231999
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:154819136 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGATGTTTTTCTTAAGAGAACT[C/G]AAAGTTGGAAAAGGCACAAGTATAT
Phenotype
MIM: 612059
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAXDC2 PubMed Links

Gene Details

Gene
FAXDC2
Gene Name
fatty acid hydroxylase domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032385.4 2030 UTR 3 NP_115761.2
XM_005268358.2 2030 UTR 3 XP_005268415.1
XM_006714753.2 2030 UTR 3 XP_006714816.1
XM_017008965.1 2030 UTR 3 XP_016864454.1
XM_017008966.1 2030 UTR 3 XP_016864455.1
Gene
LARP1
Gene Name
La ribonucleoprotein domain family member 1
There are no transcripts associated with this gene.

View Full Product Details