Product Details
- SNP ID
-
rs80343014
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:48148483 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAAGACTGCTGTTTCCTTTGATGA[A/G]GGGAAAGAGGTCCAGTCCCTTCCAA
- Phenotype
-
MIM: 615053
MIM: 610681
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ASB8
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76181949] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ASB8
- Gene Name
- ankyrin repeat and SOCS box containing 8
- Gene
- PFKM
- Gene Name
- phosphofructokinase, muscle
There are no transcripts associated with this gene.
View Full Product Details