Product Details

SNP ID
rs78184050
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:54950958 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAAAATTTTTTTCCATTTTTTTTT[C/T]CCTTTAATCTCAGAGAAATATCTGT
Phenotype
MIM: 615664
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TESPA1 PubMed Links

Gene Details

Gene
TESPA1
Gene Name
thymocyte expressed, positive selection associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098815.2 Intron NP_001092285.1
NM_001136030.2 Intron NP_001129502.1
NM_001261844.1 Intron NP_001248773.1
NM_014796.2 Intron NP_055611.1
XM_005269247.2 Intron XP_005269304.1
XM_006719715.3 Intron XP_006719778.1
XM_011539035.2 Intron XP_011537337.1
XM_011539036.1 Intron XP_011537338.1
XM_011539037.2 Intron XP_011537339.1
XM_017020262.1 Intron XP_016875751.1
XM_017020263.1 Intron XP_016875752.1

View Full Product Details