Product Details
- SNP ID
-
rs78871887
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:56273744 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTCAGAGCAAACTCTCGCTGACAG[A/G]TATATCGCGGATCAGTCTTCCTTAG
- Phenotype
-
MIM: 118950
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
COQ10A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs17118439] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- COQ10A
- Gene Name
- coenzyme Q10A
There are no transcripts associated with this gene.
- Gene
- CS
- Gene Name
- citrate synthase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004077.2 |
|
Intron |
|
|
NP_004068.2 |
View Full Product Details