Product Details

SNP ID
rs78849854
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:91051827 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGACGTCACTTGTTTTCTAAATTT[A/G]CAGGTAAGATAAAGCACATTTAGAT
Phenotype
MIM: 603288
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KERA PubMed Links

Gene Details

Gene
KERA
Gene Name
keratocan
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007035.3 Intron NP_008966.1

View Full Product Details