Product Details

SNP ID
rs76685580
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:94153925 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGCAGCCTATTGACCTCAAAATC[C/T]GCAGTCTTCTTGTGATGTTTATTTC
Phenotype
MIM: 604259
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLXNC1 PubMed Links

Gene Details

Gene
PLXNC1
Gene Name
plexin C1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005761.2 Intron NP_005752.1
XM_006719186.3 Intron XP_006719249.1
XM_011537730.2 Intron XP_011536032.1
XM_011537731.2 Intron XP_011536033.1
XM_017018671.1 Intron XP_016874160.1
XM_017018672.1 Intron XP_016874161.1

View Full Product Details