Product Details

SNP ID
rs77579203
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:103953415 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGAGCTGGACAACTAAAAGCACC[G/T]GTAGCAGTTCTCCCCAACACAGCAC
Phenotype
MIM: 191175
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C12orf73 PubMed Links

Gene Details

Gene
C12orf73
Gene Name
chromosome 12 open reading frame 73
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135570.1 1119 Intron NP_001129042.1
XM_011538718.2 1119 Missense Mutation CAG,CCG Q107P XP_011537020.1
XM_017019916.1 1119 Intron XP_016875405.1
XM_017019917.1 1119 Intron XP_016875406.1
Gene
HSP90B1
Gene Name
heat shock protein 90 beta family member 1
There are no transcripts associated with this gene.

View Full Product Details