Product Details

SNP ID
rs78988299
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:108522762 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTTGACATGGTATTAAACAAAAC[C/T]AAAGGGCTGAAACTCATGTTTAGAC
Phenotype
MIM: 611684
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FICD PubMed Links

Gene Details

Gene
FICD
Gene Name
FIC domain containing
There are no transcripts associated with this gene.

Gene
SART3
Gene Name
squamous cell carcinoma antigen recognized by T-cells 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014706.3 3357 UTR 3 NP_055521.1
XM_005269241.4 3357 UTR 3 XP_005269298.1
XM_011539026.2 3357 UTR 3 XP_011537328.1

View Full Product Details