Product Details

SNP ID
rs78064568
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:95241410 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGTATTTTGACCACAATTACCCC[A/G]CCCCTCTCTTGCCATGCTTACCTCC
Phenotype
MIM: 605900
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PDLIM1 PubMed Links

Gene Details

Gene
PDLIM1
Gene Name
PDZ and LIM domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020992.3 Intron NP_066272.1
XM_011540330.1 Intron XP_011538632.1

View Full Product Details