Product Details

SNP ID
rs74846711
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:98385556 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTTGGGTGAGACCAGCCCCAGG[C/T]TGAGAAGGGAGGCAGCATGGGACAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR1287 PubMed Links

Gene Details

Gene
MIR1287
Gene Name
microRNA 1287
There are no transcripts associated with this gene.

Gene
PYROXD2
Gene Name
pyridine nucleotide-disulphide oxidoreductase domain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032709.2 Intron NP_116098.2
XM_011540293.2 Intron XP_011538595.1
XM_011540301.2 Intron XP_011538603.1
XM_017016835.1 Intron XP_016872324.1
XM_017016836.1 Intron XP_016872325.1
XM_017016837.1 Intron XP_016872326.1
XM_017016838.1 Intron XP_016872327.1
XM_017016839.1 Intron XP_016872328.1
XM_017016840.1 Intron XP_016872329.1
XM_017016841.1 Intron XP_016872330.1

View Full Product Details