Product Details

SNP ID
rs78834718
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:99610937 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACCATGCGATGGCTGTGGAGGGG[A/G]TCTGGTAAATTACTCTGGCCTGCAC
Phenotype
MIM: 609767
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC25A28 PubMed Links

Gene Details

Gene
SLC25A28
Gene Name
solute carrier family 25 member 28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031212.3 1022 Missense Mutation ACC,ATC T336I NP_112489.3
XM_005270208.1 1022 Missense Mutation ACC,ATC T237I XP_005270265.1
XM_005270211.4 1022 Missense Mutation ACC,ATC T237I XP_005270268.1
XM_006718005.3 1022 Missense Mutation ACC,ATC T237I XP_006718068.1
XM_006718006.3 1022 Intron XP_006718069.1
XM_011540239.2 1022 Missense Mutation ACC,ATC T246I XP_011538541.1
XM_011540240.1 1022 Missense Mutation ACC,ATC T237I XP_011538542.1
XM_011540244.2 1022 Intron XP_011538546.1
XM_017016741.1 1022 Missense Mutation ACC,ATC T237I XP_016872230.1
XM_017016742.1 1022 Missense Mutation ACC,ATC T181I XP_016872231.1

View Full Product Details