Product Details

SNP ID
rs76453795
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:101826204 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAATGGGGTTGGAACCTGGGTCC[C/T]ATGGTCCCTGCCCTCTAGGTGTGCT
Phenotype
MIM: 604661 MIM: 604039
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCNIP2 PubMed Links

Gene Details

Gene
KCNIP2
Gene Name
potassium voltage-gated channel interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014591.4 1989 UTR 3 NP_055406.2
NM_173191.2 1989 UTR 3 NP_775283.1
NM_173192.2 1989 UTR 3 NP_775284.1
NM_173193.2 1989 UTR 3 NP_775285.1
NM_173194.2 1989 UTR 3 NP_775286.1
NM_173195.2 1989 UTR 3 NP_775287.1
NM_173197.2 1989 Intron NP_775289.1
XM_005269729.2 1989 UTR 3 XP_005269786.1
XM_005269730.2 1989 UTR 3 XP_005269787.1
XM_006717812.2 1989 UTR 3 XP_006717875.1
XM_011539731.2 1989 UTR 3 XP_011538033.1
XM_017016161.1 1989 UTR 3 XP_016871650.1
Gene
KCNIP2-AS1
Gene Name
KCNIP2 antisense RNA 1
There are no transcripts associated with this gene.

Gene
MGEA5
Gene Name
meningioma expressed antigen 5 (hyaluronidase)
There are no transcripts associated with this gene.

View Full Product Details