Product Details

SNP ID
rs77331202
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:116591718 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAGGCCCGAACAGCAGTGACACC[C/T]CAGAGCACACCTTGAGAGCAAATCC
Phenotype
MIM: 604422
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PNLIPRP1 PubMed Links

Gene Details

Gene
PNLIPRP1
Gene Name
pancreatic lipase related protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303135.1 Intron NP_001290064.1
NM_006229.3 Intron NP_006220.1
XM_011539868.1 Intron XP_011538170.1
XM_011539869.1 Intron XP_011538171.1

View Full Product Details